missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PNLIP Protein
A DNA sequence encoding the Mus musculus (Mouse) PNLIP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7762-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_081201.2 | |
PNLIP Protein | |
Research Use Only | |
Pnlip | |
Recombinant Protein | |
E. coli | |
REVCFDKLGC FSDDAPWSGT LDRPLKALPW SPAQINTRFL LYTNENPDNY QLITSDASNI RNSNFRTNRK TRIIIHGFID KGEENWLSDM CKNMFRVESV NCICVDWKGG SRTTYTQATQ NVRVVGAEVA LLVNVLQSDL GYSLNNVHLI GHSLGSHIAG EAGKRTFGAI GRITGLDPAE PYFQGTPEEV RLDPTDAQFV DAIHTDAGPI IPNLGFGMSQ TVGHLDFFPN GGIEMPGCQK NILSQIVDID GIWEGTRNFA ACNHLRSYKF YTDSIVNPTG FAGFSCSSYS LFTANKCFPC GSGGCPQMGH YADRYPGKTS RLYQTFYLNT GDKSNFARWR YQVTVTLSGQ KVTGHILVSL FGNGGNSKQY EVFKGSLQPG TSHVNEFDSD VDVGDLQKVK FIWYNNVINP TLPKVGASRI TVERNDGRVF NFCSQETVRE DVLLTLSPC | |
Greater than 90% as determined by SDS-PAGE. |
69060 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |