missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Phosphatidylcholine translocator ABCB4 Protein

A DNA sequence encoding the Mus musculus (Mouse) Phosphatidylcholine translocator ABCB4, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5601-ye-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916359

  • 2826.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 12-04-2019
Añadir a la cesta

Especificación

Especificación

NP_032856.2
Phosphatidylcholine translocator ABCB4 Protein
1 mg
Not determined.
Cytoplasmic Domain
Mouse
His
Tris-based buffer, 50% glycerol
18670
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Abcb4
Recombinant Protein
Yeast
DAFANARGAA YVIFDIIDNN PKIDSFSERG HKPDNIKGNL EFSDVHFSYP SRANIKILKG LNLKVKSGQT VALVGNSGCG KSTTVQLLQR LYDPTEGKIS IDGQDIRNFN VRCLREIIGV VSQEPVLFST TIAENIRYGR GNVTMDEIEK AVKEANAYDF IMKLPQKFDT LVGDRGAQLS GGQKQRIAIA RALVRNPKIL LLDEATSALD TESEAEVQAA LDKAREGRTT IVIAHRLSTI RNADVIAGFE DGVIVEQGSH SELMKKEGIY FRLVNMQTAG SQILSEEFEV ELSDEKAAGD VAPNGWKARI FRNSTKKSLK SPHQNRLDEE TNELDANVPP VSFLKVLKLN KTEWPYF
Videos