missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PGK1 Protein
A cDNA sequence encoding the PGK1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10815-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PGK1 Protein | |
Research Use Only | |
Specific activity: > 500 units/mg. One unit will convert 1 umole of 1,3-Bisphosphoglycerate to 3-PGA per minute at pH 8.0 at 37C. | |
Mouse | |
His | |
PGK1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSLSNK LTLDKLDVKG KRVVMRVDFN VPMKNNQITN NQRIKAAVPS IKFCLDNGAK SVVLMSHLGR PDGVPMPDKY SLEPVAAELK SLLGKDVLFL KDCVGPEVEN ACANPAAGTV ILLENLRFHV EEEGKGKDAS GNKVKAEPAK IDAFRASLSK LGDVYVNDAF GTAHRAHSSM VGVNLPQKAG GFLMKKELNY FAKALESPER PFLAILGGAK VADKIQLINN MLDKVNEMII GGGMAFTFLK VLNNMEIGTS LYDEEGAKIV KDLMSKAEKN GVKITLPVDF VTADKFDENA KTGQATVASG IPAGWMGLDC GTESSKKYAE AVGRAKQIVW NGPVGVFEWE AFARGTKSLM DEVVKATSRG CITIIGGGDT ATCCAKWNTE DKVSHVSTGG GASLELLEGK VLPGVDALSN V | |
Greater than 90% as determined by SDS-PAGE. |