missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse PGF1 Protein

A cDNA sequence encoding the PGF1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13028-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15932289

  • 161.00€ / 10 microgramos


Fecha estimada de envío
24-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

PGF1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
PGF1 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 0.1mM PMSF and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
AGNNSTEVEV VPFNEVWGRS YCRPMEKLVY ILDEYPDEVS HIFSPSCVLL SRCSGCCGDE GLHCVPIKTA NITMQILKIP PNRDPHFYVE MTFSQDVLCE CRPILETTKA ERRKTKGKRK RSRNSQTEEP HPHHHHHH
Videos
Documentos