missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Pcsk5 Protein

A DNA sequence encoding the Mus musculus (Mouse) Pcsk5, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6474-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053560

  • 352.00€ / 50 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001177412.1
Pcsk5 Protein
50 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
18552
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Pcsk5
Recombinant Protein
E. coli
DYDLSHAQST YFNDPKWPSM WYMHCSDNTH PCQSDMNIEG AWKRGYTGKN IVVTILDDGI ERTHPDLMQN YDALASCDVN GNDLDPMPRY DASNENKHGT RCAGEVAATA NNSHCTVGIA FNAKIGGVRM LDGDVTDMVE AKSVSYNPQH VHIYSASWGP DDDGKTVDGP APLTRQAFEN GVRMGRRGLG SVFVWASGNG GRSKDHCSCD GYTNSIYTIS ISSTAESGKK PWYLEECSST LATTYSSGES YDKKIITTDL RQRCTDNHTG TSASAPMAAG IIALALEANP FLTWRDVQHV IVRTSRAGHL NANDWKTNAA GFKVSHLYGF GLMDAE
Videos
Documentos