missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse NT5E (CD73) Protein

A cDNA sequence encoding the NT5E (CD73) was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10799-1ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915208

  • 103.00€ / 1 microgramo


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NT5E (CD73) Protein
1 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
WELTILHTND VHSRLEQTSD DSTKCLNASL CVGGVARLFT KVQQIRKEEP NVLFLDAGDQ YQGTIWFTVY KGLEVAHFMN ILGYDAMALG NHEFDNGVEG LIDPLLRNVK FPILSANIKA RGPLAHQISG LFLPSKVLSV GGEVVGIVGY TSKETPFLSN PGTNLVFEDE ISALQPEVDK LKTLNVNKII ALGHSGFEMD KLIAQKVRGV DIVVGGHSNT FLYTGNPPSK EVPAGKYPFI VTADDGRQVP VVQAYAFGKY LGYLKVEFDD KGNVITSYGN PILLNSSIPE DATIKADINQ WRIKLDNYST QELGRTIVYL DGSTQTCRFR ECNMGNLICD AMINNNLRHP DEMFWNHVSM CIVNGGGIRS PIDEKNNGTI TWENLAAVLP FGGTFDLVQL KGSTLKKAFE HSVHRYGQST GEFLQVGGIH VVYDINRKPW NRVVQLEVLC TKCRVPIYEP LEMDKVYKVT LPSYLANGGD GFQMIKDELL KHDSGDQDIS VVSEYISKMK VVYPAVEGRI KFSLEHHHHH H
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Specific activity is > 8,000 pmol/min/ug, and is defined as the amount of enzyme that hydrolyze 1.0 pmole of Adenosine 5-monophosphate to phosphate per minute per minute at pH 7.5 at 25C.
Mouse
His
NT5E protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Videos