Learn More
R&D Systems™ Recombinant Mouse NPDC-1 Fc Chimera Protein
Product Code. 18789526
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. Quantity Unit Size
18789526 50 μg 50µg
1 options
This item is not returnable. View return policy
Product Code. 18789526 Supplier R&D Systems™ Supplier No. 10970NP050
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Recombinant Protein
Measured by its ability to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons. Recombinant Mouse NPDC-1 Fc Chimera, immobilized at 2.5 μg/mL on a 96 well plate, is able to significantly enhance neurite outgrowth.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q64322.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 56654
Molecular Weight (g/mol) MolecularWeight-observed: 54-62 kDa, under reducing conditions.; MolecularWeight-theroretical: 43 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse NPDC-1 protein Mouse NPDC-1 (Ala32-Thr190, Thr33Leu) (N-terminus) Accession # Q64322.2 IEGRMD Human IgG1 Fc (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CAB-, CAB1, CAB-1, CAB1CAB, DKFZp586J0523, neural proliferation differentiation and control protein 1, neural proliferation, differentiation and control, 1, NPDC1, NPDC-1
Conjugate Unconjugated
Reconstitution Reconstitute at 1 mg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein NPDC-1