Learn More
enQuireBio™ Recombinant Mouse Nidogen-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nidogen-1, was expressed in the hosts and tags indicated.
Marca:  enQuireBio™ QP6420-ec-100ug
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 16093180
  • 770.00€ / 100 microgramos
Fecha estimada de envío: 29-05-2024
para ver el stock
Especificaciones
NP_035047.2
Nidogen-1 Protein
Research Use Only
Nid1
Recombinant Protein
E. coli
SPQRVNGKVK GRIFVGSSQV PVVFENTDLH SYVVMNHGRS YTAISTIPET VGYSLLPLAP IGGIIGWMFA VEQDGFKNGF SITGGEFTRQ AEVTFLGHPG KLVLKQQFSG IDEHGHLTIS TELEGRVPQI PYGASVHIEP YTELYHYSSS VITSSSTREY TVMEPDQDGA APSHTHIYQW RQTITFQECA HDDARPALPS TQQLSVDSVF VLYNKEERIL RYALSNSIGP VRDGSPDA
Greater than 90% as determined by SDS-PAGE.
18073
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol