missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MIP-3 beta / CCL19 Protein
A DNA sequence encoding the Mouse MIP-3 beta / CCL19, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10273-ec-20ug
Weitere Details : Peso : 0.01000kg
Spezifikation
NP_036018.1 | |
MIP-3 beta / CCL19 Protein | |
Research Use Only | |
CCL19 | |
The biological activity determined by a chemotaxis bioassay using human mature dendritic cells is in a concentration range of 10-100 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
100039053 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GANDAEDCCL SVTQRPIPGN IVKAFRYLLN EDGCRVPAVV FTTLRGYQLC APPDQPWVDR IIRRLKKSSA KNKGNSTRRS PVS | |
Greater than 97% as determined by SDS-PAGE and HPLC. |