missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MIF / GLIF Protein

A DNA sequence encoding the Mouse MIF / GLIF, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP1493-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15925509

  • 114.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 04-04-2019
Añadir a la cesta

Especificación

Especificación

NP_034928.1
MIF / GLIF Protein
10 μg
Less than 1.0 EU/ug as determined by LAL method.
Recombinant Protein
E. coli
MPMFIVNTNV PRASVPEGFL SELTQQLAQA TGKPAQYIAV HVVPDQLMTF SGTNDPCALC SLHSIGKIGG AQNRNYSKLL CGLLSDRLHI SPDRVYINYY DMNAANVGWN GSTFA
17319
Greater than 96% as determined by SDS-PAGE and HPLC.
Research Use Only
MIF
Mouse
Untagged
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized
Videos