missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MCP-3 / CCL7 Protein

A DNA sequence encoding the Mouse MCP-3 / CCL7, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10267-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923298

Cantidad Precio
1 2370.00€ / 250 microgramos
3 2211.45€ / 250 microgramos
Fecha estimada de envío
02-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_038682.1
MCP-3 / CCL7 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
QPDGPNASTC CYVKKQKIPK RNLKSYRRIT SSRCPWEAVI FKTKKGMEVC AEAHQKWVEE AIAYLDMKTP TPKP
20306
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL7
The biologically active determined by a chemotaxis bioassay using human monocytes is in a concentration range of 100-300 ng/ml.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos