missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Lymphocyte antigen 6E Protein

A DNA sequence encoding the Mus musculus (Mouse) Lymphocyte antigen 6E, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9800-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16058751

  • 342.00€ / 50 microgramos


Fecha estimada de envío
18-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001157508.1
Lymphocyte antigen 6E Protein
50 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
17069
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LY6E
Recombinant Protein
Yeast
LMCFSCTDQK NNINCLWPVS CQEKDHYCIT LSAAAGFGNV NLGYTLNKGC SPICPSENVN LNLGVASVNS YCCQSSFCNF SA
Videos
Documentos