missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Lingo1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Lingo1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8168-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053021

  • 1857.00€ / 1 miligramo


Fecha estimada de envío
18-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001298005.1
Lingo1 Protein
1 mg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
235402
Greater than 80% as determined by SDS-PAGE.
Research Use Only
Lingo1
Recombinant Protein
E. coli
PPRCECSAQD RAVLCHRKRF VAVPEGIPTE TRLLDLGKNR IKTLNQDEFA SFPHLEELEL NENIVSAVEP GAFNNLFNLR TLGLRSNRLK LIPLGVFTGL SNLTKLDISE NKIVILLDYM FQDLYNLKSL EVGDNDLVYI SHRAFSGLNS LEQLTLEKCN LTSIPTEALS HLHGLIVLRL RHLNINAIRD YSFKRLYRLK VLEISHWPYL DTMTPNCLYG LNLTSLSITH CNLTAVPYLA VRHLVYLRFL NLSYNPIGTI EGSMLHELLR LQEIQLVGGQ LAVVEPYAFR GLNYLRVLNV SGNQLTTLEE SAFHSVGNLE TLILDSNPLA CDCRLLWVFR RRWRLNFNRQ QPTCATPEFV QGKEFKDFPD VLLPNYFTCR RAHIRDRKAQ QVFVDEGHTV QFVCRADGDP PPAILWLSPR KHLVSAKSNG RLTVFPDGTL EVRYAQVQDN GTYLCIAANA GGNDSMPAHL HVRSYSPDWP HQPNKTFAFI SNQPGEGEAN STRATVPFPF DIKTLIIAT
Videos
Documentos