missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse L-FABP / FABP1 Protein

A DNA sequence encoding the Mouse L-FABP / FABP1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10197-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933058

  • 668.00€ / 100 microgramos


Fecha estimada de envío
16-10-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_059095.1
L-FABP / FABP1 Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MNFSGKYQLQ SQENFEPFMK AIGLPEDLIQ KGKDIKGVSE IVHEGKKIKL TITYGPKVVR NEFTLGEECE LETMTGEKVK AVVKLEGDNK MVTTFKGIKS VTELNGDTIT NTMTLGDIVY KRVSKRI
14080
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
Fabp1
The binding affinity of rMuFABP1 for the synthetic ligand cis-parinaric acid has been measured by fluorescence titration. Half maximal fluorescence of 2.5 uM rMuFABP1 is achieved with approximately 5 uM cis-paranaric acid.
Mouse
Untagged
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized
Videos
Documentos