missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL6 / Interleukin-6 Protein
Recombinant Protein
Marca: enQuireBio™ QP1487-100 ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001300983.1 | |
IL6 / Interleukin-6 Protein | |
Research Use Only | |
IL6 | |
Recombinant Protein | |
E. coli | |
MFPTSQVRRG DFTEDTTPNR PVYTTSQVGG LITHVLWEIV EMRKELCNGN SDCMNNDDAL AENNLKLPEI QRNDGCYQTG YNQEICLLKI SSGLLEYHSY LEYMKNNLKD NKKDKARVLQ RDTETLIHIF NQEVKDLHKI VLPTPISNAL LTDKLESQKE WLRTKTIQFI LKSLEEFLKV TLRSTRQT | |
Greater than 95% as determined by SDS-PAGE. |
16193 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 1 ng/mL in the 7TD1 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^6 units/mg. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |