missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL1F5 / IL36RN Protein

A DNA sequence encoding the Mouse IL1F5 / IL36RN, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10438-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923858

Cantidad Precio
1 267.00€ / 10 microgramos
6 249.00€ / 10 microgramos
Fecha estimada de envío
15-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_001139559.1
IL1F5 / IL36RN Protein
10 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
VLSGALCFRM KDSALKVLYL HNNQLLAGGL HAEKVIKGEE ISVVPNRALD ASLSPVILGV QGGSQCLSCG TEKGPILKLE PVNIMELYLG AKESKSFTFY RRDMGLTSSF ESAAYPGWFL CTSPEADQPV RLTQIPEDPA WDAPITDFYF QQCD
54450
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
IL36RN
Measured by its ability to inhibit IL-36gamma induced IL-6 production by human PBMCs.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos