missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL17RB Protein

A cDNA sequence encoding the IL17RB was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12410-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15920339

  • 103.00€ / 2 microgramos


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

192199
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Rspo1
Mouse
His
IL17RB protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
IL17RB Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Insect
ADPREPTIQC GSETGPSPEW MVQHTLTPGD LRDLQVELVK TSVAAEEFSI LMNISWILRA DASIRLLKAT KICVSGKNNM NSYSCVRCNY TEAFQSQTRP SGGKWTFSYV GFPVELSTLY LISAHNIPNA NMNEDSPSLS VNFTSPGCLN HVMKYKKQCT EAGSLWDPDI TACKKNEKMV EVNFTTNPLG NRYTILIQRD TTLGFSRVLE NKLMRTSVAI PVTEESEGAV VQLTPYLHTC GNDCIRREGT VVLCSETSAP IPPDDNRRML GGVEPKSCDK THTCPPCPAP ELLGGPSVFL FPPKPKDTLM ISRTPEVTCV VVDVSHEDPE VKFNWYVDGV EVHNAKTKPR EEQYNSTYRV VSVLTVLHQD WLNGKEYKCK VSNKALPAPI EKTISKAKGQ PREPQVYTLP PSRDELTKNQ VSLTCLVKGF YPSDIAVEWE SNGQPENNYK TTPPVLDSDG SFFLYSKLTV DKSRWQQGNV FSCSVMHEAL HNHYTQKSLS LSPGKHHHHH H
Videos