missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL17F / IL-17F Protein

Recombinant Protein

Marca:  enQuireBio™ QP1479-5 ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15935479

  • 84.15€ / 5 microgramos


Fecha estimada de envío
18-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_665855.2
IL17F / IL-17F Protein
5 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Mouse IL17F / IL-17F induces  IL-6 production in 3T3 cells (typical ED50 is ? 1 ug/ml).
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
257630
Greater than 95% as determined by SDS-PAGE.
Research Use Only
IL17F
Recombinant Protein
E. coli
MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAA
Videos
Documentos