missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL16 / Interleukin-16 Protein
Recombinant Protein
Marca: enQuireBio™ QP5456-1mg
Detalles adicionales : Peso : 0.01000kg
Specifications
NP_034681.2 | |
IL16 / Interleukin-16 Protein | |
Research Use Only | |
IL16 | |
Recombinant Protein | |
E. coli | |
MHDLNSSTDS AASASAASDI SVESKEATVC TVTLEKTSAG LGFSLEGGKG SLHGDKPLTI NRIFKGDRTG EMVQPGDEIL QLAGTAVQGL TRFEAWNVIK ALPDGPVTIV IRRTSLQCKQ TTASADS | |
Greater than 95% as determined by SDS-PAGE. |
16170 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse IL16 / Interleukin-16 induces primary human T cell chemotaxis (significanty different from control at Mouse IL16 / Interleukin-16 concentrations greater than 5 ng/ml). | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |