missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL13 / ALRH Protein

A DNA sequence encoding the Mouse IL13 / ALRH, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10460-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923938

Cantidad Precio
1 1910.00€ / 100 microgramos
3 1782.03€ / 100 microgramos
Fecha estimada de envío
15-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_032381.1
IL13 / ALRH Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F
16163
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
IL13
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 4 ng/ml, corresponding to a specific activity of > 2.5 M 105 IU/mg.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos