Learn More
enQuireBio™ Recombinant Mouse IL10 / Interleukin-10 Protein
Recombinant Protein
Marca:  enQuireBio™ QP5490-1mg
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15916239
  • 5607.00€ / 1 miligramo
Fecha de envío estimada
del proveedor 09-04-2019
Añadir a la cesta
Especificación
Especificación
NP_034678.1
IL10 / Interleukin-10 Protein
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 5 ng/mL in the MC/9 cell proliferation assay. The bioactivity acceptance criteria is set at 2.0 x 10^5 units/mg.
Mouse
Untagged
10 mM sodium phosphate, pH 7.5
16153
Greater than 95% as determined by SDS-PAGE.
Research Use Only
IL10
Recombinant Protein
E. coli
MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGEKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLQDQGVYKA MNEFDIFINC IEAYMMIKMK S
Videos