missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-9 / Interleukin-9 Protein
A DNA sequence encoding the Mouse IL-9 / Interleukin-9, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10457-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_032399.1 | |
IL-9 / Interleukin-9 Protein | |
Research Use Only | |
IL9 | |
The ED50 as determined by a cell proliferation assay using murine TS1 cells is less than 0.02 ng/ml, corresponding to a specific activity of > 5.0 M 107 IU/mg. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
16198 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
QRCSTTWGIR DTNYLIENLK DDPPSKCSCS GNVTSCLCLS VPTDDCTTPC YREGLLQLTN ATQKSRLLPV FHRVKRIVEV LKNITCPSFS CEKPCNQTMA GNTLSFLKSL LGTFQKTEMQ RQKSRP | |
Greater than 97% as determined by SDS-PAGE and HPLC. |