missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-3 / Interleukin-3 Protein
A DNA sequence encoding the Mouse IL-3 / Interleukin-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10452-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_034686.2 | |
IL-3 / Interleukin-3 Protein | |
Research Use Only | |
IL3 | |
The ED50 as determined by the dose-dependent stimulation of the proliferation of murine M-NFS-60 cells is less than 0.05 ng/ml, corresponding to a specific activity of > 2 M 107 IU/mg. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
16187 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
DTHRLTRTLN CSSIVKEIIG KLPEPELKTD DEGPSLRNKS FRRVNLSKFV ESQGEVDPED RYVIKSNLQK LNCCLPTSAN DSALPGVFIR DLDDFRKKLR FYMVHLNDLE TVLTSRPPQP ASGSVSPNRG TVEC | |
Greater than 98% as determined by SDS-PAGE and HPLC. |