missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL-3 / Interleukin-3 Protein

A DNA sequence encoding the Mouse IL-3 / Interleukin-3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10452-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913908

  • 299.00€ / 10 microgramos


Fecha estimada de envío
23-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_034686.2
IL-3 / Interleukin-3 Protein
10 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
DTHRLTRTLN CSSIVKEIIG KLPEPELKTD DEGPSLRNKS FRRVNLSKFV ESQGEVDPED RYVIKSNLQK LNCCLPTSAN DSALPGVFIR DLDDFRKKLR FYMVHLNDLE TVLTSRPPQP ASGSVSPNRG TVEC
16187
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
IL3
The ED50 as determined by the dose-dependent stimulation of the proliferation of murine M-NFS-60 cells is less than 0.05 ng/ml, corresponding to a specific activity of > 2 M 107 IU/mg.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos