missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL 20 Protein
A cDNA sequence encoding the IL 20 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10701-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
58181 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The biological activity was determined by measuring the dose dependent activation of STAT3 in human epidermal keratinocytes (HEK2a cells). Significant STAT3 activation is observed with >100ng/mL of Interleukin-20 recombinant mouse, corresponding to a specific activity of 10,000 units/mg of Interleukin-20 recombinant mouse. | |
Mouse | |
Untagged | |
Lyophilized from a concentrated (1 mg/ml) solution containing no additives. |
IL 20 Protein | |
Research Use Only | |
Il20 | |
Recombinant Protein | |
E. coli | |
MKGFGLAFGL FSAVGFLLWT PLTGLKTLHL GSCVITANLQ AIQKEFSEIR DSVQAEDTNI DIRILRTTES LKDIKSLDRC CFLRHLVRFY LDRVFKVYQT PDHHTLRKIS SLANSFLIIK KDLSVCHSHM ACHCGEEAME KYNQILSHFI ELELQAAVVK ALGELGILLR WMEEML | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |