missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL 18 Protein
A cDNA sequence encoding the IL 18 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12394-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
16173 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MNFGRLHCTT AVIRNINDQV LFVDKRQPVF EDMTDIDQSA SEPQTRLIIY MYKDSEVRGL AVTLSVKDSK MSTLSCKNKI ISFEEMDPPE NIDDIQSDLI FFQKRVPGHN KMEFESSLYE GHFLACQKED DAFKLILKKK DENGDKSVMF TLTNLHQS | |
Greater than 90.0% as determined by SDS-PAGE. |
IL 18 Protein | |
Research Use Only | |
Il18 | |
Mouse | |
Untagged | |
The IL 18 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4). |