missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-1 beta / IL1B Protein
A DNA sequence encoding the Mouse IL-1 beta / IL1B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10449-ec-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_032387.1 | |
IL-1 beta / IL1B Protein | |
Research Use Only | |
IL1B | |
The ED50 as determined by a cell proliferation assay using murine D10S cells is less than 2 pg/ml, corresponding to a specific activity of > 5.0 M 108 IU/mg. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
16176 | |
2 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
VPIRQLHYRL RDEQQKSLVL SDPYELKALH LNGQNINQQV IFSMSFVQGE PSNDKIPVAL GLKGKNLYLS CVMKDGTPTL QLESVDPKQY PKKKMEKRFV FNKIEVKSKV EFESAEFPNW YISTSQAEHK PVFLGNNSGQ DIIDFTMESV SS | |
Greater than 96% as determined by SDS-PAGE and HPLC. |