missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hydroxyacid oxidase 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Hydroxyacid oxidase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8261-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_034533.1 | |
Hydroxyacid oxidase 1 Protein | |
Research Use Only | |
Hao1 | |
Recombinant Protein | |
E. coli | |
MLPRLVCISD YEQHVRSVLQ KSVYDYYRSG ANDQETLADN IQAFSRWKLY PRMLRNVADI DLSTSVLGQR VSMPICVGAT AMQCMAHVDG ELATVRACQT MGTGMMLSSW ATSSIEEVAE AGPEALRWMQ LYIYKDREIS RQIVKRAEKQ GYKAIFVTVD TPYLGNRIDD VRNRFKLPPQ LRMKNFETND LAFSPKGNFG DNSGLAEYVA QAIDPSLSWD DITWLRRLTS LPIVVKGILR GDDAKEAVKH GVDGILVSNH GARQLDGVPA TIDVLPEIVE AVEGKVEVFL DGGVRKGTDV LKALALGAKA VFVGRPIIWG LAFQGEKGVQ DVLEILKEEF RLAMALSGCQ NVKVIDKTLV RKNPLAVSKI | |
Greater than 90% as determined by SDS-PAGE. |
15112 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |