missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse HB-EGF Protein
A cDNA sequence encoding the HB-EGF was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10662-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HB-EGF Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
DLEGTDLNLF KVAFSSKPQG LATPSKERNG KKKKKGKGLG KKRDPCLRKY KDYCIHGECR YLQEFRTPSC KCLPGYHGHR CHGLTL | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Mouse | |
Untagged | |
The protein was filtered (0.2microm) and lyophilized from a concentrated solution containing 10mM PB and 500mM NaCl, pH 7.4. |