missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GRK2 / ADRBK1 Protein

A DNA sequence encoding the Mus musculus (Mouse) GRK2 / ADRBK1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8014-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052161

  • 1245.00€ / 200 microgramos


Fecha estimada de envío
25-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001277747.1
GRK2 / ADRBK1 Protein
200 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
110355
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Adrbk1
Recombinant Protein
E. coli
MADLEAVLAD VSYLMAMEKS KATPAARASK KILLPEPSIR SVMQKYLEDR GEVTFEKIFS QKLGYLLFRD FCLNHLEEAK PLVEFYEEIK KYEKLETEEE RVVRSREIFD SYIMKELLAC SHPFSKNATE HVQGHLVKKQ VPPDLFQPYI EEICQNLRGD VFQKFIESDK FTRFCQWKNV ELNIHLTMND FSVHRIIGRG GFGEVYGCRK ADTGKMYAMK CLDKKRIKMK QGETLALNER IMLSLVSTGD CPFIVCMSYA FHTPDKLSFI LDLMNGGDLH YHLSQHGVFS EADMRFYAAE IILGLEHMHN RFVVYRDLKP ANILLDEHGH VRISDLGLAC DFSKKRPHAS VGTHGYMAPE VLQKGVAYDS SADWFSLGCM LFKLLRGHSP FRQHKTKDKH EIDRMTLTMA VELPDSFSPE LRSLLEGLLQ RDVNRRLGCL GRGAQEVKES PFFRSLDWQM VFLQKYPPPL IPPRGEVNAA DAFDIGSFDE EDTKGIKLLD SDQELYRNFP LTISERWQQE VAETVFDTIN AETDRLEARK KAKNKQLGHE EDYALGKDCI VHGYMSKMGN PFLTQWQRRY FYLFPNRLEW RGEGEAPQSL LTMEEIQSVE ETQIKERKCL LLKIRGGKQF VLQCDSDPEL VQWKKELRDA YREAQQLVQR VPKMKNKPRS PVVELSKVPL IQRGSANGL
Videos
Documentos