missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Glutaminyl cyclase / QPCT Protein
A DNA sequence encoding the Mus musculus (Mouse) Glutaminyl cyclase / QPCT, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6567-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001303658.1 | |
Glutaminyl cyclase / QPCT Protein | |
Research Use Only | |
QPCT | |
Recombinant Protein | |
E. coli | |
AWTQEKNHHQ PAHLNSSSLQ QVAEGTSISE MWQNDLRPLL IERYPGSPGS YSARQHIMQR IQRLQAEWVV EVDTFLSRTP YGYRSFSNII STLNPEAKRH LVLACHYDSK YFPRWDSRVF VGATDSAVPC AMMLELARAL DKKLHSLKDV SGSKPDLSLR LIFFDGEEAF HHWSPQDSLY GSRHLAQKMA SSPHPPGSRG TNQLDGMDLL VLLDLIGAAN PTFPNFFPKT TRWFNRLQAI EKELYELGLL KDHSLERKYF QNFGYGNIIQ DDHIPFLRKG VPVLHLIASP FPEVWHTMDD NEENLHASTI DNLNKIIQVF VLEYLHL | |
Greater than 90% as determined by SDS-PAGE. |
70536 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |