missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Glutaminyl cyclase / QPCT Protein

A DNA sequence encoding the Mus musculus (Mouse) Glutaminyl cyclase / QPCT, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6567-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054220

  • 2645.00€ / 1 miligramo


Fecha estimada de envío
30-06-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001303658.1
Glutaminyl cyclase / QPCT Protein
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
70536
Greater than 90% as determined by SDS-PAGE.
Research Use Only
QPCT
Recombinant Protein
E. coli
AWTQEKNHHQ PAHLNSSSLQ QVAEGTSISE MWQNDLRPLL IERYPGSPGS YSARQHIMQR IQRLQAEWVV EVDTFLSRTP YGYRSFSNII STLNPEAKRH LVLACHYDSK YFPRWDSRVF VGATDSAVPC AMMLELARAL DKKLHSLKDV SGSKPDLSLR LIFFDGEEAF HHWSPQDSLY GSRHLAQKMA SSPHPPGSRG TNQLDGMDLL VLLDLIGAAN PTFPNFFPKT TRWFNRLQAI EKELYELGLL KDHSLERKYF QNFGYGNIIQ DDHIPFLRKG VPVLHLIASP FPEVWHTMDD NEENLHASTI DNLNKIIQVF VLEYLHL
Videos
Documentos