Learn More
enQuireBio™ Recombinant Mouse Glucose-6-phosphate isomerase Protein
A DNA sequence encoding the Mus musculus (Mouse) Glucose-6-phosphate isomerase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9161-ye-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_032181.2 | |
Glucose-6-phosphate isomerase Protein | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
14751 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Gpi | |
Recombinant Protein | |
Yeast | |
AALTRNPQFQ KLLEWHRANS ANLKLRELFE ADPERFNNFS LNLNTNHGHI LVDYSKNLVN KEVMQMLVEL AKSRGVEAAR DNMFSGSKIN YTENRAVLHV ALRNRSNTPI KVDGKDVMPE VNRVLDKMKS FCQRVRSGDW KGYTGKSITD IINIGIGGSD LGPLMVTEAL KPYSKGGPRV WFVSNIDGTH IAKTLASLSP ETSLFIIASK TFTTQETITN AETAKEWFLE AAKDPSAVAK HFVALSTNTA KVKEFGIDPQ NMFEFWDWVG GRYSLWSAIG LSIALHVGFD HFEQLLSGAH WMDQHFLKTP LEKNAPVLLA LLGIWYINCY GCETHALLPY DQYMHRFAAY FQQGDMESNG KYITKSGARV DHQTGPIVWG EPGTNGQHAF YQLIHQGTKM IPCDFLIPVQ TQHPIRKGLH HKILLANFLA QTEALMKGKL PEEARKELQA AGKSPEDLEK LLPHKVFEGN RPTNSIVFTK LTPFILGALI AMYEHKIFVQ GIMWDINSFD QWGVELGKQL AKKIEPELEG SSAVTSHDSS TNGLISFIKQ QRDTKLE |