missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GLRX Protein
A cDNA sequence encoding the GLRX was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10917-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GLRX Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAQEFVN CKIQSGKVVV FIKPTCPYCR KTQEILSQLP FKQGLLEFVD ITATNNTSAI QDYLQQLTGA RTVPRVFIGK DCIGGCSDLI SMQQTGELMT RLKQIGALQL | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
GLRX protein solution (0.5 mg/ml) containing 20mM MES (pH5.0), 20% Glycerol and 0.1M NaCl. |