missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GFAP Protein
A DNA sequence encoding the Mus musculus (Mouse) GFAP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6079-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001124492.1 | |
GFAP Protein | |
Research Use Only | |
GFAP | |
Recombinant Protein | |
Yeast | |
MERRRITSAR RSYASETVVR GLGPSRQLGT MPRFSLSRMT PPLPARVDFS LAGALNAGFK ETRASERAEM MELNDRFASY IEKVRFLEQQ NKALAAELNQ LRAKEPTKLA DVYQAELREL RLRLDQLTAN SARLEVERDN FAQDLGTLRQ KLQDETNLRL EAENNLAAYR QEADEATLAR VDLERKVESL EEEIQFLRKI YEEEVRELRE QLAQQQVHVE MDVAKPDLTA ALREIRTQYE AVATSNMQET EEWYRSKFAD LTDAASRNAE LLRQAKHEAN DYRRQLQALT CDLESLRGTN ESLERQMREQ EERHARESAS YQEALARLEE EGQSLKEEMA RHLQEYQDLL NVKLALDIEI ATYRKLLEGE ENRITIPVQT FSNLQIRETS LDTKSVSEGH LKRNIVVKTV EMRDGEVIKD SKQEHKDVVM | |
Greater than 80% as determined by SDS-PAGE. |
14580 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |