missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GAD65 / GAD2 Protein

A DNA sequence encoding the Mus musculus (Mouse) GAD65 / GAD2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6062-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919709

  • 276.00€ / 10 microgramos


Fecha estimada de envío
18-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_032104.2
GAD65 / GAD2 Protein
10 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
14417
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Gad2
Recombinant Protein
E. coli
MASPGSGFWS FGSEDGSADP ENPGTARAWC QVAQKFTGGI GNKLCALLYG DSGKPAEGGG SVTSRAATGK VACTCDQKPC NCPKGDVNYA FLHATDLLPA CDGERPTLAF LQDVMNILLQ YVVKSFDRST KVIDFHYPNE LLQEYNWELA DQPQNLEEIL THCQTTLKYA IKTGHPRYFN QLSTGLDMVG LAADWLTSTA NTNMFTYEIA PVFVLLEYVT LKKMREIIGW PGGSGDGIFS PGGAISNMYA MLIARYKMFP EVKEKGMAAV PRLIAFTSEH SHFSLKKGAA ALGIGTDSVI LIKCDERGKM IPSDLERRIL EVKQKGFVPF LVSATAGTTV YGAFDPLLAV ADICKKYKIW MHVDAAWGGG LLMSRKHKWK LSGVERANSV TWNPHKMMGV PLQCSALLVR EEGLMQSCNQ MHASYLFQQD KHYDLSYDTG DKALQCGRHV DVFKLWLMWR AKGTTGFEAH IDKCLELAEY LYTIIKNREG YEMVFDGKPQ HTNVCFWFVP PSLRTLEDNE ERMSRLSKVA PVIKARMMEY GTTMVSYQPL GDKVNFFRMV ISNPAATHQD IDFLIEEIER LGQDL
Videos