missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Fractalkine / CX3CL1 Protein

A DNA sequence encoding the Mouse Fractalkine / CX3CL1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10262-ec-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933278

  • 105.00€ / 5 microgramos


Fecha estimada de envío
25-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_033168.2
Fractalkine / CX3CL1 Protein
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
QHLGMTKCEI MCGKMTSRIP VALLIRYQLN QESCGKRAIV LETTQHRRFC ADPKEKWVQD AMKHLDHQAA ALTKNG
20312
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CX3CL1
The ED50 as determined by a cell proliferation assay using human peripheral blood lymphocytes (PBL) is less than 0.5 ug/ml, corresponding to a specific activity of > 2000 IU/mg.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos