missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse FGF21 / Fibroblast Growth Factor 21 Protein

A DNA sequence encoding the Mus musculus (Mouse) FGF21 / Fibroblast Growth Factor 21, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6044-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919569

  • 2826.00€ / 1 miligramo


Fecha estimada de envío
22-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_064397.1
FGF21 / Fibroblast Growth Factor 21 Protein
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
56636
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FGF21
Recombinant Protein
E. coli
AYPIPDSSPL LQFGGQVRQR YLYTDDDQDT EAHLEIREDG TVVGAAHRSP ESLLELKALK PGVIQILGVK ASRFLCQQPD GALYGSPHFD PEACSFRELL LEDGYNVYQS EAHGLPLRLP QKDSPNQDAT SWGPVRFLPM PGLLHEPQDQ AGFLPPEPPD VGSSDPLSMV EPLQGRSPSY AS
Videos