missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse FGF21 / Fibroblast Growth Factor 21 Protein
A DNA sequence encoding the Mus musculus (Mouse) FGF21 / Fibroblast Growth Factor 21, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6044-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_064397.1 | |
FGF21 / Fibroblast Growth Factor 21 Protein | |
Research Use Only | |
FGF21 | |
Recombinant Protein | |
E. coli | |
AYPIPDSSPL LQFGGQVRQR YLYTDDDQDT EAHLEIREDG TVVGAAHRSP ESLLELKALK PGVIQILGVK ASRFLCQQPD GALYGSPHFD PEACSFRELL LEDGYNVYQS EAHGLPLRLP QKDSPNQDAT SWGPVRFLPM PGLLHEPQDQ AGFLPPEPPD VGSSDPLSMV EPLQGRSPSY AS | |
Greater than 90% as determined by SDS-PAGE. |
56636 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |