missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse ENO1 / Enolase 1 / alpha-enolase Protein

A DNA sequence encoding the Mus musculus (Mouse) ENO1 / Enolase 1 / alpha-enolase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5984-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919149

  • 368.00€ / 50 microgramos


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001020559.1
ENO1 / Enolase 1 / alpha-enolase Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
13806
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ENO1
Recombinant Protein
Yeast
SILRIHAREI FDSRGNPTVE VDLYTAKGLF RAAVPSGAST GIYEALELRD NDKTRFMGKG VSQAVEHINK TIAPALVSKK VNVVEQEKID KLMIEMDGTE NKSKFGANAI LGVSLAVCKA GAVEKGVPLY RHIADLAGNP EVILPVPAFN VINGGSHAGN KLAMQEFMIL PVGASSFREA MRIGAEVYHN LKNVIKEKYG KDATNVGDEG GFAPNILENK EALELLKTAI AKAGYTDQVV IGMDVAASEF YRSGKYDLDF KSPDDPSRYI TPDQLADLYK SFVQNYPVVS IEDPFDQDDW GAWQKFTASA GIQVVGDDLT VTNPKRIAKA ASEKSCNCLL LKVNQIGSVT ESLQACKLAQ SNGWGVMVSH RSGETEDTFI ADLVVGLCTG QIKTGAPCRS ERLAKYNQIL RIEEELGSKA KFAGRSFRNP LA
Videos