missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse DPP4 / CD26 Protein
A DNA sequence encoding the Mus musculus (Mouse) DPP4 / CD26, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9975-iv-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001153015.1 | |
DPP4 / CD26 Protein | |
Research Use Only | |
Dpp4 | |
Recombinant Protein | |
E. coli | |
SKDEAAADSR RTYSLADYLK STFRVKSYSL WWVSDFEYLY KQENNILLLN AEHGNSSIFL ENSTFESFGY HSVSPDRLFV LLEYNYVKQW RHSYTASYNI YDVNKRQLIT EEKIPNNTQW ITWSPEGHKL AYVWKNDIYV KVEPHLPSHR ITSTGEENVI YNGITDWVYE EEVFGAYSAL WWSPNNTFLA YAQFNDTGVP LIEYSFYSDE SLQYPKTVWI PYPKAGAVNP TVKFFIVNID SLSSSSSAAP IQIPAPASVA RGDHYLCDVV WATEERISLQ WLRRIQNYSV MAICDYDKIN LTWNCPSEQQ HVEMSTTGWV GRFRPAEPHF TSDGSSFYKI ISDKDGYKHI CHFPKDKKDC TFITKGAWEV ISIEALTSDY LYYISNQYKE MPGGRNLYKI QLTDHTNVKC LSCDLNPERC QYYAVSFSKE AKYYQLGCWG PGLPLYTLHR STDHKELRVL EDNSALDRML QDVQMPSKKL DFIVLNETRF WYQMILPPHF DKSKKYPLLL DVYAGPCSQK ADASFRLNWA TYLASTENII VASFDGRGSG YQGDKIMHAI NRRLGTLEVE DQIEAARQFV KMGFVDSKRV AIWGWSYGGY VTSMVLGSGS GVFKCGIAVA PVSRWEYYDS VYTERYMGLP IPEDNLDHYR NSTVMSRAEH FKQVEYLLIH GTADDNVHFQ QSAQISKALV DAGVDFQAMW YTDEDHGIAS STAHQHIYSH MSHFLQQCFS LH | |
Greater than 85% as determined by SDS-PAGE. |
13482 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |