missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Desert hedgehog Protein

A DNA sequence encoding the Mouse Desert hedgehog protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10196-ec-25ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913058

  • en la oferta / 25 microgramos

Este ítem no se puede comprar online porque su precio está sujeto a variación. Por favor, contacte con nuestro equipo de soporte de producto en el 902 239 303 para recibir ayuda.


Especificaciones

Especificaciones

NP_031883.1
Desert hedgehog Protein
25 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
IIGPGRGPVG RRRYVRKQLV PLLYKQFVPS MPERTLGASG PAEGRVTRGS ERFRDLVPNY NPDIIFKDEE NSGADRLMTE RCKERVNALA IAVMNMWPGV RLRVTEGWDE DGHHAQDSLH YEGRALDITT SDRDRNKYGL LARLAVEAGF DWVYYESRNH IHVSVKADNS LAVRAGG
13363
Greater than 98% as determined by SDS.­PAGE and HPLC.
Research Use Only
DHH
The ED50 as determined by its ability to induce alkaline phosphatase production by murine MC3T3­E1 cells is 5­20 ug/ml.
Mouse
Untagged
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized
Videos