missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Decorin / DCN / SLRR1B Protein
A DNA sequence encoding the Mus musculus (Mouse) Decorin / DCN / SLRR1B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5915-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001177380.1 | |
Decorin / DCN / SLRR1B Protein | |
Research Use Only | |
DCN | |
Recombinant Protein | |
Yeast | |
GIIPYDPDNP LISMCPYRCQ CHLRVVQCSD LGLDKVPWDF PPDTTLLDLQ NNKITEIKEG AFKNLKDLHT LILVNNKISK ISPEAFKPLV KLERLYLSKN QLKELPEKMP RTLQELRVHE NEITKLRKSD FNGLNNVLVI ELGGNPLKNS GIENGAFQGL KSLSYIRISD TNITAIPQGL PTSLTEVHLD GNKITKVDAP SLKGLINLSK LGLSFNSITV MENGSLANVP HLRELHLDNN KLLRVPAGLA QHKYIQVVYL HNNNISAVGQ NDFCRAGHPS RKASYSAVSL YGNPVRYWEI FPNTFRCVYV RSAIQLGNYK | |
Greater than 90% as determined by SDS-PAGE. |
13179 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |