missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse D-dopachrome decarboxylase Protein

A DNA sequence encoding the Mus musculus (Mouse) D-dopachrome decarboxylase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5921-ye-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928699

  • 985.00€ / 200 microgramos


Fecha estimada de envío
17-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_034157.1
D-dopachrome decarboxylase Protein
200 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
13202
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Ddt
Recombinant Protein
Yeast
PFVELETNLP ASRIPAGLEN RLCAATATIL DKPEDRVSVT IRPGMTLLMN KSTEPCAHLL VSSIGVVGTA EQNRTHSASF FKFLTEELSL DQDRIVIRFF PLEAWQIGKK GTVMTFL
Videos
Documentos