missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Cytochrome P450 3A13 Protein

A DNA sequence encoding the Mus musculus (Mouse) Cytochrome P450 3A13, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7720-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16050411

  • 1715.00€ / 500 microgramos


Fecha estimada de envío
29-06-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_031845.1
Cytochrome P450 3A13 Protein
500 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
13113
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Cyp3a13
Recombinant Protein
E. coli
MDLIPNFSME TWMLLATSLV LLYLYGTHSH GIFKKLGIPG PKPLPFLGTI LAYQKGFWEC DIQCHKKYGK MWGLYDGRQP VLAITDPDII KTVLVKECYS TFTNRRRFGP VGILKKAISI SENEEWKRIR ALLSPTFTSG RLKEMFPIIN QFTDVLVRNM RQGLGEGKPT SMKDIFGAYS MDVITATSFG VNIDSLNNPQ DPFVEKIKKL LKFDIFDPLF LSVTLFPFLT PVFDALNVSL FPRDVISFFT TSVERMKENR MKEKEKQRVD FLQLMINSQN YKTKESHKAL SDVEIVAQSV IFIFAGYETT SSALSFALYL LAIHPDVQKK LQDEIDAALP NKAPATYDTL LQMEYLDMVV NETLRLYPIA GRLERVCKTD VEINGLFIPK GTVVMIPTFA LHKDPKYWPE PEEFRPERFS KKNQDSINPY MYLPFGSGPR NCIGMRFALI NMKVALVRVL QNFTVQPCKE TEIPLKLSKQ GLLQPENPLL LKVVSRDETV SDE
Videos
Documentos