missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cysteine sulfinic acid decarboxylase Protein
A DNA sequence encoding the Mus musculus (Mouse) Cysteine sulfinic acid decarboxylase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5874-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_659191.1 | |
Cysteine sulfinic acid decarboxylase Protein | |
Research Use Only | |
Csad | |
Recombinant Protein | |
E. coli | |
MADSKPLRTL DGDPVAVEAL LQDVFGIVVD EAILKGTSAS EKVCEWKEPE ELKQLLDLEL QSQGESREQI LERCRTVIHY SVKTGHPRFF NQLFSGLDPH ALAGRIITES LNTSQYTYEI APVFVLMEEE VLKKLRALVG WNSGDGVFCP GGSISNMYAM NLARFQRYPD CKQRGLRALP PLALFTSKEC HYSITKGAAF LGLGTDSVRV VKADERGRMI PEDLERQIIL AEAEGSVPFL VSATSGTTVL GAFDPLDAIA DVCQRHGLWF HVDAAWGGSV LLSRTHRHLL DGIQRADSVA WNPHKLLAAG LQCSALLLRD TSNLLKRCHG SQASYLFQQD KFYDVALDTG DKVVQCGRRV DCLKLWLMWK AQGGQGLERR IDQAFALTRY LVEEIKKREG FELVMEPEFV NVCFWFVPPS LRGKKESPDY SQRLSQVAPV LKERMVKKGT MMIGYQPHGT RANFFRMVVA NPILAQADID FLLGELELLG QDL | |
Greater than 90% as determined by SDS-PAGE. |
246277 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |