missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cyclophilin B Protein
A cDNA sequence encoding the Cyclophilin B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10569-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
19035 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 1,000 nmol/min/mg, and is defined as the amount of enzyme that cleaves 1nmole of suc-AAFP-PNA per minute at 37C in Tris€“HCl pH 8.0 using chymotrypsin. | |
Mouse | |
His | |
Cyclophilin B protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
Cyclophilin B Protein | |
Research Use Only | |
Ppib | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNDKKKG PKVTVKVYFD LQIGDESVGR VVFGLFGKTV PKTVDNFVAL ATGEKGFGYK NSKFHRVIKD FMIQGGDFTR GDGTGGKSIY GERFPDENFK LKHYGPGWVS MANAGKDTNG SQFFITTVKT SWLDGKHVVF GKVLEGMDVV RKVESTKTDSRDKPLKDVII VDSGKIEVEK PFAIAKE | |
Greater than 95.0% as determined by SDS-PAGE. |