missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CXCL5 / ENA-78 Protein

A DNA sequence encoding the Mouse CXCL5 / ENA-78, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10253-ec-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933248

  • 267.00€ / 20 microgramos


Fecha estimada de envío
31-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_033167.2
CXCL5 / ENA-78 Protein
20 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
TELRCVCLTV TPKINPKLIA NLEVIPAGPQ CPTVEVIAKL KNQKEVCLDP EAPVIKKIIQ KILGSDKKKA
20311
Greater than 96% as determined by SDS-PAGE and HPLC.
Research Use Only
CXCL5
The biological activity determined by a chemoattract bioassay using human neutrophils is in a concentration range of 10-100 ng/ml.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos