missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CXCL5 / ENA-78 Protein

A DNA sequence encoding the Mus musculus (Mouse) CXCL5 / ENA-78, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8653-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055781

  • 245.00€ / 10 microgramos


Fecha estimada de envío
04-05-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_033167.2
CXCL5 / ENA-78 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
20311
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CXCL5
Recombinant Protein
E. coli
IAATELRCVC LTVTPKINPK LIANLEVIPA GPQCPTVEVI AKLKNQKEVC LDPEAPVIKK IIQKILGSDK KK
Videos
Documentos