missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CXCL14 / BRAK Protein

A DNA sequence encoding the Mouse CXCL14 / BRAK, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10260-ec-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933268

  • 267.00€ / 20 microgramos


Fecha estimada de envío
13-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_062514.2
CXCL14 / BRAK Protein
20 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SKCKCSRKGP KIRYSDVKKL EMKPKYPHCE EKMVIVTTKS MSRYRGQEHC LHPKLQSTKR FIKWYNAWNE KRRVYEE
57266
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
Cxcl14
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 1.0-10 ng/ml.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos